Water, [3H] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT924D-500MCI MT924D-1CI MT924D-2CI MT924D-5CI
101973-984EA 1268.46 USD
101973-984 101973-986 101973-988 101973-990
Water, [3H] ≥98%, MORpure™
Water, [3H]
CAS Number: 13670-17-2
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR