2',3'-Dideoxyinosine

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: 9-((2R,5S)-5-(Hydroxymethyl)tetrahydrofuran-2-yl)-9H-purin-6-ol

MT711-50UCI MT711-250UCI
101973-470EA 1601.92 USD
101973-470 101973-472
2',3'-Dideoxyinosine
2',3'-Dideoxyinosine
Formula: C₁₀H₁₂N₄O₃
MW: 236.23 g/mol
MDL Number: MFCD00077728
CAS Number: 69655-05-6
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR