L-Leucine, [4,5-3H] ≥97%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT672EL-250UCI MT672EL-1MCI MT672EL-5MCI
101973-362EA 477.31 USD
101973-362 101973-364 101973-366
L-Leucine, [4,5-3H] ≥97%, MORpure™
L-Leucine, [4,5-3H]
CAS Number: 2087903-59-9
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR