P-Aminobenzoic acid, [3,5-3H] ≥97%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT599-250UCI MT599-1MCI
101973-104EA 2700.38 USD
101973-104 101973-106
P-Aminobenzoic acid, [3,5-3H] ≥97%, MORpure™
P-Aminobenzoic acid, [3,5-3H]
CAS Number: 117610-59-0
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR