D-Ribavirin, [triazole-5-3H] ≥97%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT540-250UCI MT540-1MCI
101973-028EA 2700.38 USD
101973-028 101973-030
D-Ribavirin, [triazole-5-3H] ≥97%, MORpure™
D-Ribavirin, [triazole-5-3H]
CAS Number: 2086327-92-4
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR