Benzoic acid, free acid, [3H] ≥97%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT1882-250UCI MT1882-1MCI MT1882-5MCI
101972-690EA 2700.38 USD
101972-690 101972-692 101972-694
Benzoic acid, free acid, [3H] ≥97%, MORpure™
Benzoic acid, free acid, [3H]
CAS Number: 2086336-82-3
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR