Fluconazole

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: 2-(2,4-Difluorophenyl)-1,3-bis(1H-1,2,4-triazol-1-yl)propan-2-ol, 2-(2,4-Difluorophenyl)-1,3-di(1H-1,2,4-triazol-1-yl)propan-2-ol

MT1752-250UCI MT1752-1MCI
101972-554EA 2700.38 USD
101972-554 101972-556
Fluconazole
Fluconazole
Formula: C₁₃H₁₂F₂N₆O
MW: 306.28 g/mol
MDL Number: MFCD00274549
CAS Number: 86386-73-4
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR