Atenolol, [ring-3H] ≥97% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT1724-250UCI MT1724-1MCI
101972-492EA 1896.15 USD
101972-492 101972-494
Atenolol, [ring-3H] ≥97% (by HPLC), MORpure™
Atenolol, [ring-3H]
CAS Number: 2086336-88-9
UN: 2910
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR