L-Asparagine, [3H] ≥97%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MT1534-250UCI MT1534-1MCI
101972-212EA 2857.31 USD
101972-212 101972-214
L-Asparagine, [3H] ≥97%, MORpure™
L-Asparagine, [3H]
CAS Number: 2089105-16-6
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR