3'-Deoxy-2',3'-didehydrothymidine, [4-14C] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC284-10UCI MC284-50UCI
101971-698EA 1693.46 USD
101971-698 101971-700
3'-Deoxy-2',3'-didehydrothymidine, [4-14C] ≥98%, MORpure™
3'-Deoxy-2',3'-didehydrothymidine, [4-14C]
CAS Number: 336806-13-4
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR