7,10,13,16,19-Docosapentaenoic acid, [1-14C] ≥98% (by HPLC), MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC2224-10UCI MC2224-50UCI
101971-218EA 3138.46 USD
101971-218 101971-220
7,10,13,16,19-Docosapentaenoic acid, [1-14C] ≥98% (by HPLC), MORpure™
7,10,13,16,19-Docosapentaenoic acid, [1-14C]
CAS Number: 288146-37-2
UN: 2910
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR