Uric acid, [8-14C] ≥95%

Supplier: MORAVEK BIOCHEMICALS MS

MC1394-50UCI MC1394-250UCI
101970-462EA 1719.62 USD
101970-462 101970-464
Uric acid, [8-14C] ≥95%
Uric acid, [8-14C]
CAS Number: 92334-71-9
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR