Benzocaine, [ring-14C] ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

MC1105-10UCI MC1105-50UCI MC1105-250UCI
101970-100EA 1693.46 USD
101970-100 101970-102 101970-104
Benzocaine, [ring-14C] ≥98%, MORpure™
Benzocaine, [ring-14C]
CAS Number: 2088103-46-0
UN: 2910

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR