Lamivudine

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: (-)-1-[(2R,5S)-2-(Hydroxymethyl)-1,3-oxathiolan-5-yl]cytosine, 4-Amino-1-((2R,5S)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl)pyrimidin-2(1H)-one, 3TC

M975-5MG M975-1MG
101970-044EA 778.08 USD
101970-044 101970-042
Lamivudine
Lamivudine
Formula: C₈H₁₁N₃O₃S
MW: 229.2600 g/mol
MDL Number: MFCD00869739
CAS Number: 134678-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR