3-(3,4-Dihydroxyphenyl)-L-alanine
Supplier: MORAVEK BIOCHEMICALS MS
Synonyms:
Dopar, L-dopa, L-β-3,4-Dihydroxyphenyl-Alanine, 3-Hydroxy-L-tyrosine, Levodopa
M1887-1MG
M1887-5MG
101969-810EA
294.23
USD
101969-810
101969-812
3-(3,4-Dihydroxyphenyl)-L-alanine
3-(3,4-Dihydroxyphenyl)-L-alanine
Formula:
C₉H₁₁NO₄ MW: 197.19 g/mol |
MDL Number:
MFCD00002598 CAS Number: 59-92-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...