Efavirenz ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: (4S)-6-Chloro-4-(2-cyclopropylethynyl)-1,4-dihydro-4-(trifluoromethyl)-2H-3,1-benzoxazin-2-one

M1821-1MG M1821-5MG
101969-720EA 294.23 USD
101969-720 101969-722
Efavirenz ≥98%, MORpure™
Efavirenz
Formula: C₁₄H₉ClF₃NO₂
MW: 315.68 g/mol
MDL Number: MFCD05662344
CAS Number: 154598-52-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR