7-Deaza-7-iodo-2'-deoxyguanosine ≥98%, MORpure™

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: 7-Iodo-7-deaza-2'-deoxyguanosine, 2-Amino-7-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-iodo-1H-pyrrolo[2,3-d]pyrimidin-4(7H)-one

M1778-1MG M1778-5MG
101969-556EA 294.23 USD
101969-556 101969-558
7-Deaza-7-iodo-2'-deoxyguanosine ≥98%, MORpure™
7-Deaza-7-iodo-2'-deoxyguanosine
Formula: C₁₁H₁₃IN₄O₄
MW: 392.1498 g/mol
MDL Number: MFCD07778651
CAS Number: 172163-62-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR