5-Bromo-2′-deoxycytidine hydrate ≥98%, MORpure™, HPLC purified

Supplier: MORAVEK BIOCHEMICALS MS

M1755-1MG M1755-5MG
101969-476EA 294.23 USD
101969-476 101969-478
5-Bromo-2′-deoxycytidine hydrate ≥98%, MORpure™, HPLC purified
5-Bromo-2′-deoxycytidine hydrate
Formula: C₉H₁₂BrN₃O₄·×H₂O
MW: 306.116 g/mol (anhy)
CAS Number: 1022-79-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR