5-Bromo-2'-deoxyuridine (Broxuridine)
Supplier: MORAVEK BIOCHEMICALS MS
Synonyms:
5-Bromo-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 5-Bromo-2'-deoxyuridine, 5-BrdU
M1753-1MG
M1753-5MG
101969-468EA
294.23
USD
101969-468
101969-470
5-Bromo-2'-deoxyuridine (Broxuridine)
5-Bromo-2'-deoxyuridine (Broxuridine)
Formula:
C₉H₁₁BrN₂O₅ MW: 307.10 g/mol |
MDL Number:
MFCD00006529 CAS Number: 59-14-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...