2',3'-Dideoxyguanosine
Supplier: MORAVEK BIOCHEMICALS MS
Synonyms:
2-Amino-9-((2R,5S)-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1H-purin-6(9H)-one
M1749-1MG
M1749-5MG
101969-456EA
294.23
USD
101969-456
101969-458
2',3'-Dideoxyguanosine
2',3'-Dideoxyguanosine
Formula:
C₁₀H₁₃N₅O₃ MW: 251.24 g/mol |
MDL Number:
MFCD00057074 CAS Number: 85326-06-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...