2',3'-Dideoxycytidine

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: 4-Amino-1-((2R,5S)-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one, Zalcitabine

M1748-1MG M1748-5MG
101969-452EA 294.23 USD
101969-452 101969-454
2',3'-Dideoxycytidine
2',3'-Dideoxycytidine
Formula: C₉H₁₃N₃O₃
MW: 211.22 g/mol
MDL Number: MFCD00012188
CAS Number: 7481-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR