2',3'-Dideoxyadenosine

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: ((2S,5R)-5-(6-Amino-9H-purin-9-yl)tetrahydrofuran-2-yl)methanol

M1747-1MG M1747-5MG
101969-448EA 294.23 USD
101969-448 101969-450
2',3'-Dideoxyadenosine
2',3'-Dideoxyadenosine
Formula: C₁₀H₁₃N₅O₂
MW: 235.24 g/mol
MDL Number: MFCD00010534
CAS Number: 4097-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR