Ribavirin

Supplier: MORAVEK BIOCHEMICALS MS

Synonyms: 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1H-1,2,4-triazole-3-carboxamide

M1736-1MG M1736-5MG
101969-408EA 294.23 USD
101969-408 101969-410
Ribavirin
Ribavirin
Formula: C8H12N4O5
MW: 244.2047 g/mol
MDL Number: MFCD00058564
CAS Number: 36791-04-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR