Anti-4-1BB (CD137) Rat Monoclonal Antibody [clone: LOB12.3]

Supplier: Bio X Cell
BP0169-5MG BP0169-25MG BP0169-100MG BP0169-50MG
103479-718EA 5100 USD
103479-718 103479-720 103479-722 103479-724 76554-958 76555-272 76556-774 76557-328
Anti-4-1BB (CD137) Rat Monoclonal Antibody [clone: LOB12.3]
Antibodies
InVivoPlus anti-mouse 4-1BB (CD137), Clone: LOB12.3, Isotype: rat IgG2a, Applications: in vivo activation of 4-1BB

Type: Primary
Antigen: 4-1BB (CD137)
Clonality: Monoclonal
Clone: LOB12.3
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR