Anti-HER2 (neu) Mouse Monoclonal Antibody [clone: 7.16.4]

Supplier: Bio X Cell
BE0277-25MG BE0277-100MG BE0277-50MG BE0277-1MG BE0277-5MG
103254-392EA 3920 USD
103254-392 103254-394 103254-396 103254-398 103572-926 76555-218 76555-750 76555-898 76556-138 76556-180
Anti-HER2 (neu) Mouse Monoclonal Antibody [clone: 7.16.4]
Antibodies
InVivoMab anti-human/rat HER2 (neu), Clone: 7.16.4, Isotype: mouse IgG2a, Applications: in vivo and in vitro HER2/neu inhibition, IP, IF, FC

Type: Primary
Antigen: HER2 (neu)
Clonality: Monoclonal
Clone: 7.16.4
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR