Anti-CD8 alpha Rat Monoclonal Antibody [clone: 53-6.7]

Supplier: Bio X Cell
BP0004-1-5MG BP0004-1-50MG BP0004-1-25MG BP0004-1-100MG
103254-280EA 5100 USD
103254-280 103254-282 103254-284 103254-286 76555-592 76555-650 76556-140 76556-672
Anti-CD8 alpha Rat Monoclonal Antibody [clone: 53-6.7]
Antibodies
InVivoPlus anti-mouse CD8α, Clone: 53-6.7, Isotype: rat IgG2a, κ, Applications: in vivo CD8+ T cell depletion, Immunofluorescence, Flow cytometry

Type: Primary
Antigen: CD8alpha
Clonality: Monoclonal
Clone: 53-6.7
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR