Anti-CD3 epsilon Armenian Hamster Monoclonal Antibody [clone: 145-2C11]

Supplier: Bio X Cell
BP0001-1-100MG BP0001-1-5MG BP0001-1-50MG BP0001-1-25MG
103254-264EA 5100 USD
103254-264 103254-266 103254-268 103254-270 76555-680 76555-808 76555-854 76556-392
Anti-CD3 epsilon Armenian Hamster Monoclonal Antibody [clone: 145-2C11]
Antibodies
InVivoPlus anti-mouse CD3ε, Clone: 145-2C11, Isotype: Armenian hamster IgG, Applications: in vivo T cell depletion, in vitro T cell stimulation/activation, Immunofluorescence, Flow cytometry

Type: Primary
Antigen: CD3epsilon
Clonality: Monoclonal
Clone: 145-2C11
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR