Anti-IgG1 Mouse Monoclonal Antibody [clone: RG11/39.4]

Supplier: Bio X Cell
BE0250-100MG BE0250-5MG BE0250-50MG BE0250-1MG BE0250-25MG
103254-144EA 3920 USD
103254-144 103254-146 103254-148 103254-150 103572-618 76554-482 76554-486 76556-602 76556-762 76556-946
Anti-IgG1 Mouse Monoclonal Antibody [clone: RG11/39.4]
Antibodies
InVivoMab anti-rat IgG1, Clone: RG11/39.4, Isotype: mouse IgG2a, κ, Applications: Flow cytometry

Type: Primary
Antigen: IgG1
Clonality: Monoclonal
Clone: RG11/39.4
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR