Anti-IFNgamma Mouse Monoclonal Antibody [clone: B27]

Supplier: Bio X Cell
BE0245-50MG BE0245-5MG BE0245-1MG BE0245-100MG BE0245-25MG
103254-104EA 3920 USD
103254-104 103254-106 103254-108 103254-110 103572-608 76554-900 76555-772 76556-224 76556-770 76557-286
Anti-IFNgamma Mouse Monoclonal Antibody [clone: B27]
Antibodies
InVivoMab anti-human IFNγ, Clone: B27, Isotype: mouse IgG1, Applications: Flow cytometry

Type: Primary
Antigen: IFNgamma
Clonality: Monoclonal
Clone: B27
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR