Anti-FcRn heavy chain heterodimers Mouse Monoclonal Antibody [clone: 1G3]

Supplier: Bio X Cell
BE0222-1MG BE0222-25MG BE0222-50MG BE0222-100MG BE0222-5MG
103254-040EA 3920 USD
103254-040 103254-042 103254-044 103254-046 103572-570 76554-848 76555-550 76556-380 76556-484 76556-570
Anti-FcRn heavy chain heterodimers Mouse Monoclonal Antibody [clone: 1G3]
Antibodies
InVivoMab anti-rat FcRn heavy chain heterodimers, Clone: 1G3, Isotype: mouse IgG1, Applications: Immunofluorescence, ELISA, Flow cytometry

Type: Primary
Antigen: FcRn heavy chain heterodimers
Clonality: Monoclonal
Clone: 1G3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR