Anti-KLRG-1 Syrian Hamster Monoclonal Antibody [clone: 2F1]

Supplier: Bio X Cell
BE0201-100MG BE0201-50MG BE0201-1MG BE0201-25MG BE0201-5MG
103253-904EA 3920 USD
103253-904 103253-906 103253-908 103253-910 103572-908 76554-932 76555-316 76556-072 76556-418 76556-752
Anti-KLRG-1 Syrian Hamster Monoclonal Antibody [clone: 2F1]
Antibodies
InVivoMab anti-mouse/human KLRG-1, Clone: 2F1, Isotype: Syrian hamster IgG, Applications: Flow cytometry

Type: Primary
Antigen: KLRG-1
Clonality: Monoclonal
Clone: 2F1
Conjugation:
Epitope:
Host: Syrian Hamster
Isotype: IgG
Reactivity: Mouse, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR