Anti-Vgamma2 TCR Armenian Hamster Monoclonal Antibody [clone: UC3-10A]

Supplier: Bio X Cell
BE0168-100MG BE0168-1MG BE0168-25MG BE0168-50MG BE0168-5MG
103253-872EA 3920 USD
103253-872 103253-874 103253-876 103253-878 103572-858 76556-114 76556-162 76556-366 76556-620 76557-020
Anti-Vgamma2 TCR Armenian Hamster Monoclonal Antibody [clone: UC3-10A]
Antibodies
InVivoMab anti-mouse Vγ2 TCR, Clone: UC3-10A, Isotype: Armenian hamster IgG, κ, Applications: in vivo γδ T cell depletion, Flow cytometry

Type: Primary
Antigen: Vgamma2 TCR
Clonality: Monoclonal
Clone: UC3-10A
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR