Anti-2C TCR Mouse Monoclonal Antibody [clone: 1B2]

Supplier: Bio X Cell
BE0069-50MG BE0069-25MG BE0069-1MG BE0069-5MG BE0069-100MG
103253-630EA 3920 USD
103253-630 103253-632 103253-634 103253-636 103572-758 76554-544 76554-842 76555-266 76556-754 76556-924
Anti-2C TCR Mouse Monoclonal Antibody [clone: 1B2]
Antibodies
InVivoMab anti-mouse 2C TCR, Clone: 1B2, Isotype: mouse IgG1, Applications: Immunofluorescence, Flow cytometry

Type: Primary
Antigen: 2C TCR
Clonality: Monoclonal
Clone: 1B2
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR