Anti-IL-7 Mouse Monoclonal Antibody [clone: M25]

Supplier: Bio X Cell
BE0048-50MG BE0048-25MG BE0048-5MG BE0048-1MG BE0048-100MG
103253-486EA 3920 USD
103253-486 103253-488 103253-490 103253-492 103572-490 76554-356 76554-408 76554-804 76556-696 76557-034
Anti-IL-7 Mouse Monoclonal Antibody [clone: M25]
Antibodies
InVivoMab anti-mouse/human IL-7, Clone: M25, Isotype: mouse IgG2b, Applications: in vivo IL-7 neutralization, in vivo IL-7 receptor stimulation (as a complex with IL-7)

Type: Primary
Antigen: IL-7
Clonality: Monoclonal
Clone: M25
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Mouse, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR