Anti-IL-2 Rat Monoclonal Antibody [clone: S4B6-1]

Supplier: Bio X Cell
BE0043-1-5MG BE0043-1-25MG BE0043-1-100MG BE0043-1-50MG BE0043-1-1MG
103253-446EA 3920 USD
103253-446 103253-448 103253-450 103253-452 103572-480 76555-818 76555-890 76556-292 76556-866 76557-330
Anti-IL-2 Rat Monoclonal Antibody [clone: S4B6-1]
Antibodies
InVivoMab anti-mouse IL-2, Clone: S4B6-1, Isotype: rat IgG2a, Applications: in vivo IL-2 neutralization, in vivo IL-2 receptor stimulation (as a complex with IL-2)

Type: Primary
Antigen: IL-2
Clonality: Monoclonal
Clone: S4B6-1
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR