Anti-CD132 (common gamma chain) Rat Monoclonal Antibody [clone: 3E12]

Supplier: Bio X Cell
BE0271-100MG BE0271-5MG BE0271-1MG BE0271-50MG BE0271-25MG
103253-408EA 3920 USD
103253-408 103253-410 103253-412 103253-414 103572-642 76554-360 76554-370 76554-714 76556-854 76556-864
Anti-CD132 (common gamma chain) Rat Monoclonal Antibody [clone: 3E12]
Antibodies
InVivoMab anti-mouse CD132 (common γ chain), Clone: 3E12, Isotype: rat IgG2b, Applications: in vivo γc blockade, Functional assays, Immunoprecipitation, Flow cytometry

Type: Primary
Antigen: CD132 (common gamma chain)
Clonality: Monoclonal
Clone: 3E12
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR