Anti-OVA Mouse Monoclonal Antibody [clone: F2-3.58]

Supplier: Bio X Cell
RT0267-25MG RT0267-1MG
103253-368EA 150 USD
103253-368 103253-370 103253-372 103253-374 76554-654 76557-350
Anti-OVA Mouse Monoclonal Antibody [clone: F2-3.58]
Antibodies
ReadyTag anti-OVA, Clone: F2-3.58, Isotype: mouse IgG1, Applications: ELISA

Type: Primary
Antigen: OVA
Clonality: Monoclonal
Clone: F2-3.58
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR