Anti-GST Mouse Monoclonal Antibody [clone: F50-3D12.2]

Supplier: Bio X Cell
RT0264-25MG RT0264-1MG
103253-344EA 150 USD
103253-344 103253-346 103253-348 103253-350 76554-644 76556-834
Anti-GST Mouse Monoclonal Antibody [clone: F50-3D12.2]
Antibodies
ReadyTag anti-GST, Clone: F50-3D12.2, Isotype: mouse IgG1, Applications: Western blot

Type: Primary
Antigen: GST
Clonality: Monoclonal
Clone: F50-3D12.2
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR