Anti-IFNgamma Rat Monoclonal Antibody [clone: XMG1.2]

Supplier: Bio X Cell
BP0055-50MG BP0055-25MG BP0055-5MG BP0055-100MG
103253-264EA 5100 USD
103253-264 103253-266 103253-268 103253-270 76554-488 76556-310 76556-852 76557-072
Anti-IFNgamma Rat Monoclonal Antibody [clone: XMG1.2]
Antibodies
InVivoPlus anti-mouse IFNγ, Clone: XMG1.2, Isotype: rat IgG1, κ, Applications: in vivo IFNγ neutralization, in vitro IFNγ neutralization, ELISPOT, Flow cytometry

Type: Primary
Antigen: IFNgamma
Clonality: Monoclonal
Clone: XMG1.2
Conjugation:
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR