Anti-Ly6G/Ly6C (Gr-1) Rat Monoclonal Antibody [clone: RB6-8C5]

Supplier: Bio X Cell
BP0075-5MG BP0075-100MG BP0075-50MG BP0075-25MG
103253-240EA 5100 USD
103253-240 103253-242 103253-244 103253-246 76554-752 76555-700 76555-864 76556-232
Anti-Ly6G/Ly6C (Gr-1) Rat Monoclonal Antibody [clone: RB6-8C5]
Antibodies
InVivoPlus anti-mouse Ly6G/Ly6C (Gr-1) , Clone: RB6-8C5, Isotype: rat IgG2b, κ, Applications: in vivo depletion of Gr-1+ myeloid cells, Flow cytometry, Immunohistochemistry (paraffin), Immunohistochemistry (frozen)

Type: Primary
Antigen: Ly6G/Ly6C (Gr-1)
Clonality: Monoclonal
Clone: RB6-8C5
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR