Anti-TCR Vgamma1.1/Cr4 Armenian Hamster Monoclonal Antibody [clone: 2.11]

Supplier: Bio X Cell
BE0257-25MG BE0257-100MG BE0257-5MG BE0257-50MG BE0257-1MG
103253-184EA 3920 USD
103253-184 103253-186 103253-188 103253-190 103572-628 76554-728 76555-632 76556-096 76556-468 76556-706
Anti-TCR Vgamma1.1/Cr4 Armenian Hamster Monoclonal Antibody [clone: 2.11]
Antibodies
InVivoMab anti-mouse TCR Vγ1.1/Cr4, Clone: 2.11, Isotype: Armenian hamster IgG, Applications: Flow cytometry

Type: Primary
Antigen: TCR Vgamma1.1/Cr4
Clonality: Monoclonal
Clone: 2.11
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR