Anti-IFNgamma Mouse Monoclonal Antibody [clone: B133.5]

Supplier: Bio X Cell
BE0235-1MG BE0235-5MG BE0235-50MG BE0235-25MG BE0235-100MG
103253-160EA 3920 USD
103253-160 103253-162 103253-164 103253-166 103572-590 76555-950 76556-116 76556-296 76556-684 76557-048
Anti-IFNgamma Mouse Monoclonal Antibody [clone: B133.5]
Antibodies
InVivoMab anti-human IFNγ, Clone: B133.5, Isotype: mouse IgG1, Applications: in vitro IFNγ neutralization

Type: Primary
Antigen: IFNgamma
Clonality: Monoclonal
Clone: B133.5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR