Anti-CD3 Mouse Monoclonal Antibody [clone: UCHT1 (Leu-4)(T3)]

Supplier: Bio X Cell
BE0231-1MG BE0231-50MG BE0231-25MG BE0231-5MG BE0231-100MG
103253-128EA 3920 USD
103253-128 103253-130 103253-132 103253-134 103572-582 76554-342 76555-126 76556-690 76556-820 76557-082
Anti-CD3 Mouse Monoclonal Antibody [clone: UCHT1 (Leu-4)(T3)]
Antibodies
InVivoMab anti-human CD3, Clone: UCHT1 (Leu-4)(T3), Isotype: mouse IgG1, κ, Applications: in vivo T cell depletion in humanized mice, ex vivo T cell inhibition for xenographs, Flow cytometry

Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: UCHT1 (Leu-4)(T3)
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR