Anti-CCL2 (MCP-1) Armenian Hamster Monoclonal Antibody [clone: 2H5]

Supplier: Bio X Cell
BE0185-50MG BE0185-5MG BE0185-1MG BE0185-100MG BE0185-25MG
103252-984EA 3920 USD
103252-984 103252-986 103252-988 103252-990 103572-880 76555-108 76555-788 76555-838 76556-922 76557-086
Anti-CCL2 (MCP-1) Armenian Hamster Monoclonal Antibody [clone: 2H5]
Antibodies
InVivoMab anti-human/mouse, rat CCL2 (MCP-1), Clone: 2H5, Isotype: Armenian hamster IgG, κ, Applications: in vivo CCL2 neutralization, Immunohistochemistry (frozen)

Type: Primary
Antigen: CCL2 (MCP-1)
Clonality: Monoclonal
Clone: 2H5
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR