Anti-CD1d (CD1.1) Rat Monoclonal Antibody [clone: 20H2 (HB-323)]

Supplier: Bio X Cell
BE0179-100MG BE0179-25MG BE0179-5MG BE0179-50MG BE0179-1MG
103252-936EA 3920 USD
103252-936 103252-938 103252-940 103252-942 103572-556 76555-532 76555-638 76556-814 76557-106 76557-320
Anti-CD1d (CD1.1) Rat Monoclonal Antibody [clone: 20H2 (HB-323)]
Antibodies
InVivoMab anti-mouse CD1d (CD1.1), Clone: 20H2 (HB-323), Isotype: rat IgG1, κ, Applications: iNKT cell neutralization, in vivo CD1d blockade, Flow cytometry

Type: Primary
Antigen: CD1d (CD1.1)
Clonality: Monoclonal
Clone: 20H2 (HB-323)
Conjugation:
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR