Anti-Kappa Immunoglobulin Light Chain Rat Monoclonal Antibody [clone: 187.1/HB-58]

Supplier: Bio X Cell
BE0176-100MG BE0176-1MG BE0176-5MG BE0176-25MG BE0176-50MG
103252-920EA 3920 USD
103252-920 103252-922 103252-924 103252-926 103572-552 76554-334 76554-846 76556-414 76556-708 76556-994
Anti-Kappa Immunoglobulin Light Chain Rat Monoclonal Antibody [clone: 187.1/HB-58]
Antibodies
InVivoMab anti-mouse Kappa Immunoglobulin Light Chain, Clone: 187.1/HB-58, Isotype: rat IgG1, κ, Applications: Immunofluorescence

Type: Primary
Antigen: Kappa Immunoglobulin Light Chain
Clonality: Monoclonal
Clone: 187.1/HB-58
Conjugation:
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR