Anti-Jagged2 Armenian Hamster Monoclonal Antibody [clone: HMJ2-1]

Supplier: Bio X Cell
BE0125-25MG BE0125-5MG BE0125-100MG BE0125-50MG BE0125-1MG
103252-684EA 3920 USD
103252-684 103252-686 103252-688 103252-690 103572-818 76554-844 76554-874 76555-384 76555-410 76555-986
Anti-Jagged2 Armenian Hamster Monoclonal Antibody [clone: HMJ2-1]
Antibodies
InVivoMab anti-mouse Jagged2, Clone: HMJ2-1, Isotype: Armenian hamster IgG, Applications: in vivo Jagged 2 neutralization

Type: Primary
Antigen: Jagged2
Clonality: Monoclonal
Clone: HMJ2-1
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR