Anti-T15 VH and T15 VL regions of IgM Mouse Monoclonal Antibody [clone: AB1-2 (HB-33)]

Supplier: Bio X Cell
BE0072-1MG BE0072-100MG BE0072-5MG BE0072-50MG BE0072-25MG
103252-486EA 3920 USD
103252-486 103252-488 103252-490 103252-492 103572-764 76554-588 76555-098 76555-252 76555-340 76557-060
Anti-T15 VH and T15 VL regions of IgM Mouse Monoclonal Antibody [clone: AB1-2 (HB-33)]
Antibodies
InVivoMab anti-mouse T15 VH and T15 VL regions of IgM, Clone: AB1-2 (HB-33), Isotype: mouse IgG1, κ, Applications: ELISA

Type: Primary
Antigen: T15 VH and T15 VL regions of IgM
Clonality: Monoclonal
Clone: AB1-2 (HB-33)
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR