Rabbit IgG Isotype Control

Supplier: Bio X Cell
BE0095-5MG BE0095-50MG BE0095-100MG BE0095-1MG BE0095-25MG
103252-630EA 3920 USD
103252-630 103252-632 103252-634 103252-636 103572-508 76554-416 76556-040 76556-074 76556-396 76556-966
Rabbit IgG Isotype Control
Antibodies
InVivoMab polyclonal rabbit IgG, Clone: Polyclonal, Isotype: Rabbit IgG, Applications: in vivo administration

  • Recommended Dilution Buffer: IP0070

Recommended Dilution Buffer: IP0070

Type: Primary
Antigen:
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR