Human IgG Isotype Control

Supplier: Bio X Cell
BE0092-25MG BE0092-50MG BE0092-100MG BE0092-1MG BE0092-5MG
103252-606EA 3920 USD
103252-606 103252-608 103252-610 103252-612 103572-502 76554-458 76555-968 76556-574 76556-798 76557-004
Human IgG Isotype Control
Antibodies
InVivoMab polyclonal human IgG, Clone: Polyclonal, Isotype: human IgG, Applications: in vivo administration

Recommended Dilution Buffer: IP0070

Type: Primary
Antigen:
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Human
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR